Ammonium benzoate ≥98%

Warning

Synonyms: Benzoic acid ammonium salt

183334-1KG 183334-250G
220040-395EA 56.11 USD
220040-395 220040-396
Ammonium benzoate ≥98%
Ammonium benzoate
Formula: C₇H₉NO₂
MW: 139.15 g/mol
Melting Pt: 197.8 °C
Density: 1.26 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00013069
CAS Number: 1863-63-4
EINECS: 217-468-9
Merck Index: 12,00521

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR