3,3-Dimethyl-1-butanol ≥98%

Synonyms: 3,3-Dimethylbutan-1-ol

183105-10G 183105-50G
220040-376EA 140 USD
220040-376 220040-377
3,3-Dimethyl-1-butanol ≥98%
3,3-Dimethyl-1-butanol
Formula: C₆H₁₄O
MW: 102.18 g/mol
Boiling Pt: 142…143 °C (1013 hPa)
Melting Pt: –65…–60 °C
Density: 0.844 g/cm³ (15 °C)
Storage Temperature: Ambient
MDL Number: MFCD00002928
CAS Number: 624-95-3
EINECS: 210-872-6
UN: 1987
ADR: 3,III

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR