Dibutyltin oxide

Warning

Synonyms: Dibutyltin(IV) oxide, Dibutyloxostannane, Di-n-butyltin oxide, Di-n-butyltin(IV) oxide

183083-500G 183083-100G
220040-374EA 88.62 USD
220040-374 220040-373
Dibutyltin oxide
Dibutyltin oxide
Formula: C₈H₁₈OSn
MW: 248.94 g/mol
Melting Pt: ? 300 °C
Density: 1.6 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00001992
CAS Number: 818-08-6
EINECS: 212-449-1
UN: 3146
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR