1-Methylisatin

Synonyms: 1-Methylindoline-2,3-dione, 1-Methyl-2,3-dihydroindole-2,3-dione, N-Methylindoline-2,3-dione, N-Methylisatin, 1-Methyl-1H-indole-2,3-dione, N-Methyl-1H-indole-2,3-dione, N-Methyl-2,3-dihydroindole-2,3-dione

183075-10G 183075-1G
220040-847EA 213.23 USD
220040-847 220040-372
1-Methylisatin
1-Methylisatin
Formula: C₉H₇NO₂
MW: 161.16 g/mol
Melting Pt: 139 °C
Density: 1.4 g/cm³ (20 °C)
MDL Number: MFCD00005812
CAS Number: 2058-74-4
EINECS: 218-164-9
UN: 2811
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR