trans,trans-2,4-Hexadien-1-ol

Synonyms: (2E,4E)-Hexa-2,4-dien-1-ol, Trans,trans-2,4-hexadien-1-ol, (2E,4E)-1-Hydroxyhexa-2,4-diene, Sorbic alcohol, trans,trans-Hexa-2,4-dien-1-ol

183059-50G 183059-10G
220040-844EA 266 USD
220040-844 220040-843
trans,trans-2,4-Hexadien-1-ol
trans,trans-2,4-Hexadien-1-ol
Formula: C₆H₁₀O
MW: 98.14 g/mol
MDL Number: MFCD00002925
CAS Number: 17102-64-6
EINECS: 241-173-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR