1-Dodecyl-3-methylimidazolium iodide

18289-50G-F 18289-5G-F
220040-834EA 1160 USD
220040-834 220040-835
1-Dodecyl-3-methylimidazolium iodide
1-Dodecyl-3-methylimidazolium iodide
Formula: C₁₆H₃₁IN₂
MW: 378.34 g/mol
CAS Number: 81995-09-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR