Sodium alginate

Synonyms: Alginic acid sodium salt, Algin

180947-250G 180947-100G 180947-500G
220040-359EA 123.45 USD
220040-359 220040-358 220040-360
Sodium alginate
Sodium alginate
Storage Temperature: Ambient
MDL Number: MFCD00081310
CAS Number: 9005-38-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR