1-Ethyl-3-methylimidazolium methylsulfate

Synonyms: 1-Ethyl-3-methyl-1H-imidazol-3-ium methyl sulphate

18086-50G-F 18086-5G-F
220040-352EA 217 USD
220040-352 220040-353
1-Ethyl-3-methylimidazolium methylsulfate
1-Ethyl-3-methylimidazolium methylsulphate
Formula: C₇H₁₄N₂O₄S
MW: 222.27 g/mol
Density: 1.28 g/cm³ (20 °C)
MDL Number: MFCD08457697
CAS Number: 516474-01-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR