Diphenyl diselenide ≥98%

Danger

180629-100G 180629-5G 180629-25G
220040-698EA 426.46 USD
220040-698 220040-700 220040-699
Diphenyl diselenide ≥98%
Diphenyl diselenide
Formula: C₁₂H₁₀Se₂
MW: 312.13 g/mol
Boiling Pt: 377 °C (1013 hPa)
Melting Pt: 58 - 61 °C
Density: 1.32 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003001
CAS Number: 1666-13-3
EINECS: 216-780-2
UN: 3283
ADR: 6.1,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR