1-Amino-2,2,2-trifluoroethane hydrochloride

Synonyms: 2,2,2-Trifluoroethanamine hydrochloride, 2,2,2-Trifluoroethylamine hydrochloride

180386-50G 180386-10G
220040-677EA 286.46 USD
220040-677 220040-676
1-Amino-2,2,2-trifluoroethane hydrochloride
1-Amino-2,2,2-trifluoroethane hydrochloride
Formula: C₂H₅ClF₃N
MW: 135.52 g/mol
Melting Pt: 222…226 °C
MDL Number: MFCD00012875
CAS Number: 373-88-6
EINECS: 206-771-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR