2-Naphthoic acid

Synonyms: β-Naphthoic acid

180246-25G 180246-5G
220040-666EA 196 USD
220040-666 220040-667
2-Naphthoic acid
2-Naphthoic acid
Formula: C₁₁H₈O₂
MW: 172.18 g/mol
Boiling Pt: 333 °C (1013 hPa)
Melting Pt: 180…183 °C
Density: 1.382 g/cm³ (20 °C)
MDL Number: MFCD00004101
CAS Number: 93-09-4
EINECS: 202-217-8
Merck Index: 12,06469

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR