Potassium hydrogen phthalate 99.95-100.05% analytical standard, for titrimetry

179922-100G 179922-500G
220040-615EA 26.92 USD
220040-615 220040-616
Potassium hydrogen phthalate 99.95-100.05% analytical standard, for titrimetry
Potassium hydrogen phthalate
Formula: HOOCC₆H₄COOK
Melting Pt: 295…300 °C
Density: 1.6362 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00013070
CAS Number: 877-24-7
EINECS: 212-889-4
Merck Index: 13,07694
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4652479e822d1591ac7eb5e29f892b8c94588ad4&vid=75071da7f30b07f7bd077699a3b54ec89d80f0b8&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR