Borane dimethyl sulfide complex

Synonyms: Borane-methyl sulphide complex

179825-25ML 179825-800ML 179825-100ML
220040-607EA 53.65 USD
220040-607 220040-608 220040-605
Borane dimethyl sulfide complex
Borane dimethyl sulphide complex
Formula: C₂H₉BS
MW: 75.97 g/mol
Melting Pt: –27 °C
Density: 1.28 g/cm³ (20 °C)
MDL Number: MFCD00013189
CAS Number: 13292-87-0
EINECS: 236-313-6
UN: 2924
ADR: 3,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR