Boron trichloride 1 M in dichloromethane

178934-800ML 178934-100ML
220040-343EA 326.77 USD
220040-343 220040-342
Boron trichloride 1 M in dichloromethane
Boron trichloride
Formula: BCl₃
MW: 117.17 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00011313
CAS Number: 10294-34-5
EINECS: 233-658-4
UN: 2927
ADR: 6.1,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR