Tetrabutylammonium hydroxide 40% (w/w) in aqueous solution

Danger

178780-1L 178780-2.5L 178780-250ML 178780-50ML
220040-326EA 546.92 USD
220040-326 220040-327 220040-328 220040-329
Tetrabutylammonium hydroxide 40% (w/w) in aqueous solution
Tetrabutylammonium hydroxide
Formula: C₁₆H₃₇NO
MW: 259.48 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00009425
CAS Number: 2052-49-5
EINECS: 218-147-6
UN: 3267
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR