Isoquinoline-1-amine ≥99%

Warning

Synonyms: Isoquinolin-1-amine, 1-Aminoisoquinoline

178594-5G 178594-1G
220039-706EA 315 USD
220039-706 220039-705
Isoquinoline-1-amine ≥99%
Isoquinoline-1-amine
Formula: C₉H₈N₂
MW: 144.18 g/mol
Boiling Pt: 324 °C (1013 hPa)
Melting Pt: 122…124 °C
Density: 1.21 g/cm³ (20 °C)
MDL Number: MFCD00024137
CAS Number: 1532-84-9
EINECS: 216-243-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR