2',4'-Dichloroacetophenone ≥96%

Synonyms: 2′,4′-Dichloroacetophenone

178373-100G 178373-25G
220040-143EA 172.31 USD
220040-143 220040-144
2',4'-Dichloroacetophenone ≥96%
2',4'-Dichloroacetophenone
Formula: C₈H₆Cl₂O
MW: 189.04 g/mol
Boiling Pt: 247 °C (1013 hPa)
Melting Pt: 42 °C
Density: 1.32 g/cm³ (20 °C)
MDL Number: MFCD00000581
CAS Number: 2234-16-4
EINECS: 218-780-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR