4-Aminobutan-1-ol ≥98%

Danger

Synonyms: 4-Hydroxybutylamine, 4-Amino-1-butanol

178330-5G 178330-1G
220040-132EA 117.38 USD
220040-132 220040-131
4-Aminobutan-1-ol ≥98%
4-Aminobutan-1-ol
Formula: C₄H₁₁NO
MW: 89.14 g/mol
Boiling Pt: 206 °C (1013 hPa)
Melting Pt: 16…18 °C
Density: 0.967 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00008230
CAS Number: 13325-10-5
EINECS: 236-364-4
UN: 2735
ADR: 8,III

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR