1-Chloro-2-methylpropane ≥98%

178004-500ML 178004-100ML
220040-104EA 90.46 USD
220040-104 220040-103
1-Chloro-2-methylpropane ≥98%
1-Chloro-2-methylpropane
Formula: C₄H₉Cl
MW: 92.57 g/mol
Boiling Pt: 68 °C (1013 hPa)
Melting Pt: -131 °C
Density: 0.8829 g/cm³ (20 °C)
Flash Pt: -17 °C
Storage Temperature: Ambient
MDL Number: MFCD00000954
CAS Number: 513-36-0
EINECS: 208-157-9
UN: 1127
ADR: 3,II
Merck Index: 13,05155

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR