Isobutyl chloroformate ≥98%

Danger

Synonyms: Chloroformic acid isobutyl ester

177989-100G 177989-25G
220039-775EA 44.91 USD
220039-775 220039-776
Isobutyl chloroformate ≥98%
Isobutyl chloroformate
Formula: C₅H₉ClO₂
MW: 136.58 g/mol
Boiling Pt: 128.7 °C (1013 hPa)
Melting Pt: -80 °C
Density: 1.05 g/cm³ (20 °C)
Flash Pt: 30 °C
MDL Number: MFCD00000642
CAS Number: 543-27-1
EINECS: 208-840-1
UN: 2742
ADR: 6.1,II
Merck Index: 13,05156

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR