1,1,1-Trifluoroiodoethane ≥99%

Warning

Synonyms: 1-Iodo-2,2,2-trifluoroethane, 2,2,2-Trifluoroethyliodide, 2-Iodo-1,1,1-trifluoroethane, 2,2,2-Trifluoroiodoethane, 1,1,1-Trifluoro-2-iodoethane

177814-25G 177814-5G
220040-098EA 191.69 USD
220040-098 220040-099
1,1,1-Trifluoroiodoethane ≥99%
1,1,1-Trifluoroiodoethane
Formula: C₂H₂F₃I
MW: 209.94 g/mol
Boiling Pt: 54.7 °C (1013 hPa)
Density: 2.1499 g/cm³ (20 °C)
MDL Number: MFCD00001082
CAS Number: 353-83-3
EINECS: 206-541-0
UN: 2810
ADR: 6.1,III

Order Now

Specification Test Results

Assay 99%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR