2,5-Dimethylfuran

177717-100ML 177717-25ML
220040-080EA 147.54 USD
220040-080 220040-081
2,5-Dimethylfuran
2,5-Dimethylfuran
Formula: C₆H₈O
MW: 96.13 g/mol
Boiling Pt: 92 °C (1013 hPa)
Melting Pt: –62 °C
Density: 0.903 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003250
CAS Number: 625-86-5
EINECS: 210-914-3
UN: 3271
ADR: 3,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR