1,3-Cyclopentanedione ≥97%

Synonyms: Cyclopentane-1,3-dione

177164-5G 177164-1G
220040-053EA 129.23 USD
220040-053 220040-052
1,3-Cyclopentanedione ≥97%
1,3-Cyclopentanedione
Formula: C₅H₆O₂
MW: 98.1 g/mol
Melting Pt: 149 - 152 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00001405
CAS Number: 3859-41-4
EINECS: 223-372-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR