3-Quinolinecarboxylic acid ≥98%

177148-5G 177148-1G 177148-25G
220040-050EA 110.92 USD
220040-050 220040-048 220040-049
3-Quinolinecarboxylic acid ≥98%
3-Quinolinecarboxylic acid
Formula: C₁₀H₇NO₂
MW: 173.17 g/mol
Boiling Pt: 345 °C (1013 hPa)
Melting Pt: 282 - 283 °C
Storage Temperature: Ambient
MDL Number: MFCD00006770
CAS Number: 6480-68-8
EINECS: 229-337-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR