3-Quinolinecarbaldehyde ≥98%

Synonyms: Quinoline-3-carboxaldehyde, Quinoline-3-carbaldehyde, 3-Quinolinecarboxaldehyde

177121-5G 177121-1G
220040-047EA 125.38 USD
220040-047 220039-621
3-Quinolinecarbaldehyde ≥98%
3-Quinolinecarbaldehyde
Formula: C₁₀H₇NO
MW: 157.17 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00006768
CAS Number: 13669-42-6
EINECS: 237-147-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR