4-Quinolinecarbaldehyde ≥97%

Synonyms: Quinoline-4-carbaldehyde, Quinoline-4-carboxaldehyde, 4-Quinolinecarboxaldehyde

176966-10G 176966-2.5G
220039-614EA 247.69 USD
220039-614 220039-615
4-Quinolinecarbaldehyde ≥97%
4-Quinolinecarbaldehyde
Formula: C₁₀H₇NO
MW: 157.17 g/mol
Boiling Pt: 122…123 °C (4 mmHg)
Melting Pt: 150 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00006781
CAS Number: 4363-93-3
EINECS: 224-453-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR