Anthracene-[D₁₀]

Synonyms: Anthracene-[²H₁₀]

176591-1G 176591-5G
220040-009EA 240 USD
220040-009 220040-010
Anthracene-[D₁₀]
Anthracene-[D₁₀]
Formula: C₁₄D₁₀
MW: 188.15 g/mol
Boiling Pt: 340 °C (1013 hPa)
Storage Temperature: Refrigerator
CAS Number: 1719-06-8
EINECS: 217-004-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR