Silver trifluoromethanesulfonate ≥99%

Synonyms: Silver trifluoromethanesulphonate, Silver triflate, Trifluoromethanesulphonic acid silver salt

176435-1G 176435-10G 176435-25G
220039-535EA 26.86 USD
220039-535 220039-534 220039-536
Silver trifluoromethanesulfonate ≥99%
Silver trifluoromethanesulphonate
Formula: CAgF₃O₃S
MW: 256.94 g/mol
Melting Pt: 356 °C
MDL Number: MFCD00013226
CAS Number: 2923-28-6
EINECS: 220-882-2
UN: 3260
ADR: 8,III

Order Now

Specification Test Results

Assay 99%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR