5-Bromo-2-methylpyridine

17636-1G 17636-250MG
220039-454EA 383.08 USD
220039-454 220039-527
5-Bromo-2-methylpyridine
5-Bromo-2-methylpyridine
Formula: C₆H₆BrN
MDL Number: MFCD00661170
CAS Number: 3430-13-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR