Trifluoromethanesulfonic anhydride ≥99%

Danger

Synonyms: Triflic anhydride

176176-10G 176176-250G 176176-1KG 176176-5G 176176-50G
220039-440EA 47.55 USD
220039-440 220039-442 220039-441 220039-444 220039-443
Trifluoromethanesulfonic anhydride ≥99%
Trifluoromethanesulphonic anhydride
Formula: C₂F₆O₅S₂
MW: 282.14 g/mol
Boiling Pt: 83 °C (1013 hPa)
Density: 1.67 g/cm³ (20 °C)
Storage Temperature: Refrigerator
MDL Number: MFCD00000408
CAS Number: 358-23-6
EINECS: 206-616-8
UN: 3265
ADR: 8,II

Order Now

Specification Test Results

Assay 99%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR