Naphthalene-[D₈]

Warning

Synonyms: Naphthalene-[²H₈]

176044-10G 176044-5G 176044-1G
220039-989EA 793.85 USD
220039-989 220039-991 220039-990
Naphthalene-[D₈]
Naphthalene-[D₈]
Formula: C₁₀D₈
MW: 136.11 g/mol
Melting Pt: 81 - 83 °C
Flash Pt: 78 °C
Storage Temperature: Ambient
MDL Number: MFCD00001743
CAS Number: 1146-65-2
EINECS: 214-552-7
UN: 1334
ADR: 4.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR