p-Xylene-[D₁₀]

Danger

Synonyms: p-Xylene-[²H₁₀], 1,4-Dimethylbenzene-D10

175927-10G 175927-5G
220039-974EA 106.54 USD
220039-974 220039-975
p-Xylene-[D₁₀]
p-Xylene-[D₁₀]
Formula: C₆D₄(CD₃)₂
MW: 116.09 g/mol
Boiling Pt: 135 °C (1013 hPa)
Melting Pt: 13.3 °C
Density: 0.948 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00037699
CAS Number: 41051-88-1
EINECS: 255-193-6
UN: 1307
ADR: 3,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR