o-Xylene-[D₁₀]

Danger

Synonyms: 1,2-Dimethylbenzene-D10, o-Xylene-[²H₁₀]

175900-5G 175900-10G
220039-972EA 41.23 USD
220039-972 220039-971
o-Xylene-[D₁₀]
o-Xylene-[D₁₀]
Formula: C₆D₄(CD₃)₂
MW: 116.09 g/mol
Boiling Pt: 142 °C (1013 hPa)
Melting Pt: –25 °C
Density: 0.953 g/cm³ (20 °C)
Flash Pt: 30 °C
Storage Temperature: Ambient
MDL Number: MFCD00037698
CAS Number: 56004-61-6
EINECS: 259-942-8
UN: 1307
ADR: 3,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR