tert-Butanol-[D10]

Danger

Synonyms: Trimethylcarbinol-D10, tert-Butyl alcohol-D10, 2-Methyl-2-propanol-D10

175889-1G 175889-5G
220039-967EA 49.85 USD
220039-967 220039-968
tert-Butanol-[D10]
tert-Butanol-[D10]
Formula: D₃CC(CD₃)(OD)CD₃
MW: 84.04 g/mol
Boiling Pt: 82 °C (1013 hPa)
Storage Temperature: Ambient
CAS Number: 53001-22-2
EINECS: 258-288-0
UN: 1120
ADR: 3,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR