tert-Butanol-[D1]

Danger

Synonyms: tert-Butyl alcohol-D1, Trimethylcarbinol-D1, 2-Methyl-2-propanol-D1

175765-5G 175765-25G
220038-497EA 140.15 USD
220038-497 220039-966
tert-Butanol-[D1]
tert-Butanol-[D1]
Formula: C₄H₉DO
MW: 75.11 g/mol
Density: 0.797 g/cm³ (20 °C)
Storage Temperature: Ambient
CAS Number: 3972-25-6
EINECS: 223-597-1
UN: 1120
ADR: 3,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR