Bromobenzene-[D₅]

Synonyms: Phenylbromide-D5, Bromobenzene-[²H₅]

175730-25G 175730-5G
220039-964EA 89.85 USD
220039-964 220039-965
Bromobenzene-[D₅]
Bromobenzene-[D₅]
Formula: C₆D₅Br
MW: 161.97 g/mol
Boiling Pt: 156.6 °C (1013 hPa)
Melting Pt: -31 °C
Density: 1.55 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00000056
CAS Number: 4165-57-5
EINECS: 224-013-8
UN: 2514
ADR: 3,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR