Benzene-[D₁]

175722-5G 175722-1G
220039-963EA 251.08 USD
220039-963 220039-962
Benzene-[D₁]
Benzene-[D₁]
Formula: C₆H₅D
CAS Number: 1120-89-4
EINECS: 214-321-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR