(3-Mercaptopropyl)trimethoxysilane

Synonyms: Trimethoxy(3-mercaptopropyl)silane, 3-(Trimethoxysilyl)propane-1-thiol, 3-(Trimethoxysilyl)-1-propanethiol

175617-100G 175617-500G 175617-25G
220039-946EA 184.38 USD
220039-946 220039-948 220039-947
(3-Mercaptopropyl)trimethoxysilane
(3-Mercaptopropyl)trimethoxysilane
Formula: HSH₂CCH₂CH₂Si(OCH₃)₃
MW: 196.34 g/mol
Boiling Pt: 215 °C (1013 hPa)
Melting Pt: –50 °C
Density: 1.056 g/cm³ (20 °C)
Flash Pt: >65 °C
Storage Temperature: Ambient
MDL Number: MFCD00004901
CAS Number: 4420-74-0
EINECS: 224-588-5
UN: 3082
ADR: 9,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR