Diethoxy(dimethyl)silane

Synonyms: Dimethyldiethoxysilane

175595-500ML 175595-100ML
220039-942EA 247.15 USD
220039-942 220039-941
Diethoxy(dimethyl)silane
Diethoxy(dimethyl)silane
Formula: C₆H₁₆O₂Si
MW: 148.28 g/mol
Boiling Pt: 114 °C (1013 hPa)
Melting Pt: -87 °C
Density: 0.853 g/cm³ (20 °C)
MDL Number: MFCD00009068
CAS Number: 78-62-6
EINECS: 201-127-6
UN: 2380
ADR: 3,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR