Trichlorosilane ≥99%

Danger

175552-5G 175552-100G 175552-500G
220039-935EA 54.71 USD
220039-935 220039-933 220039-934
Trichlorosilane ≥99%
Trichlorosilane
Formula: HCl₃Si
MW: 135.45 g/mol
Boiling Pt: 32 °C (1013 hPa)
Melting Pt: -127.2 °C
Density: 1.344 g/cm³ (20 °C)
Flash Pt: -28 °C
Storage Temperature: Ambient
MDL Number: MFCD00011518
CAS Number: 10025-78-2
EINECS: 233-042-5
UN: 1295
ADR: 4.3,I
REACH: 01-2119494046-35
Merck Index: 12,09776

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR