5-Bromo-2-chlorobenzoic acid ≥98%

Warning

175544-25G 175544-100G
220039-932EA 32.31 USD
220039-932 220039-931
5-Bromo-2-chlorobenzoic acid ≥98%
5-Bromo-2-chlorobenzoic acid
Formula: C₇H₄BrClO₂
MW: 235.46 g/mol
Boiling Pt: 316 °C (1013 hPa)
Melting Pt: 157 - 160 °C
Storage Temperature: Ambient
MDL Number: MFCD00002415
CAS Number: 21739-92-4
EINECS: 244-558-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR