3-Ethylaniline ≥98%

175498-50G 175498-10G
220039-755EA 191.69 USD
220039-755 220039-754
3-Ethylaniline ≥98%
3-Ethylaniline
Formula: C₈H₁₁N
MW: 121.18 g/mol
Boiling Pt: 212 °C (1013 hPa)
Melting Pt: –8 °C
Density: 0.97 g/cm³ (20 °C)
Flash Pt: 85 °C
Storage Temperature: Ambient
MDL Number: MFCD00007818
CAS Number: 587-02-0
EINECS: 209-594-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR