4-Isopropylphenol ≥98%

Danger

175404-100G 175404-5G
220039-749EA 118.46 USD
220039-749 220039-750
4-Isopropylphenol ≥98%
4-Isopropylphenol
Formula: C₉H₁₂O
MW: 136.19 g/mol
Boiling Pt: 213 °C (1013 hPa)
Melting Pt: 60 °C
Density: 0.99 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00002372
CAS Number: 99-89-8
EINECS: 202-798-8
UN: 2430
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR