Antimony(V) fluoride ≥99%

Warning

175129-25G 175129-5G
220039-923EA 424.62 USD
220039-923 220039-924
Antimony(V) fluoride ≥99%
Antimony(V) fluoride
Formula: F₅Sb
MW: 216.75 g/mol
Boiling Pt: 149.4 °C (1013 hPa)
Melting Pt: 7 °C
Density: 2.99 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011219
CAS Number: 7783-70-2
EINECS: 232-021-8
UN: 1732
ADR: 8,II
Merck Index: 13,00702

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR