Nitrosium tetrafluoroborate

Synonyms: Nitrosyl tetrafluoroborate, Nitrosonium tetrafluoroborate

175064-25G 175064-5G 175064-50G
220039-918EA 158.54 USD
220039-918 220039-920 220039-919
Nitrosium tetrafluoroborate
Nitrosium tetrafluoroborate
Formula: BF₄NO
MW: 116.81 g/mol
MDL Number: MFCD00011433
CAS Number: 14635-75-7
EINECS: 238-679-2
UN: 3260
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR