4-Methoxy-1-naphthol

Synonyms: 4-Methoxynaphthalen-1-ol, 1-Hydroxy-4-methoxynaphthalene

174556-5G 174556-25G
220039-900EA 92.4 USD
220039-900 220039-899
4-Methoxy-1-naphthol
4-Methoxy-1-naphthol
Formula: C₁₁H₁₀O₂
MW: 174.2 g/mol
Boiling Pt: 313 °C (1013 hPa)
Melting Pt: 131 °C
MDL Number: MFCD00003976
CAS Number: 84-85-5
EINECS: 201-566-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR