2-Bromo-5-hydroxy-4-methoxybenzaldehyde, Technical Grade

17347-5G 17347-25G
220039-668EA 756.92 USD
220039-668 220039-667
2-Bromo-5-hydroxy-4-methoxybenzaldehyde, Technical Grade
2-Bromo-5-hydroxy-4-methoxybenzaldehyde
Formula: C₈H₇BrO₃
MW: 231.05 g/mol
MDL Number: MFCD00195552
CAS Number: 2973-59-3
EINECS: 426-540-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR