Tributylphosphine oxide

Warning

Synonyms: Tri-n-butylphosphineoxide, Oseltamivir impurity H

172766-25G 172766-5G
220039-846EA 124.62 USD
220039-846 220039-847
Tributylphosphine oxide
Tributylphosphine oxide
Formula: C₁₂H₂₇OP
MW: 218.32 g/mol
Boiling Pt: 348 °C (1013 hPa)
Melting Pt: 62…63 °C
MDL Number: MFCD00002082
CAS Number: 814-29-9
EINECS: 212-394-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR