Ethyl-p-toluate ≥99%

Synonyms: Ethyl 4-methylbenzoate, Ethyl-4-methylbenzoate, 4-Methylbenzoic acid ethyl ester, 4-Toluic acid ethyl ester, p-Toluic acid ethyl ester

172693-250ML 172693-50ML
220039-611EA 185.23 USD
220039-611 220039-842
Ethyl-p-toluate ≥99%
Ethyl-p-toluate
Formula: C₁₀H₁₂O₂
MW: 164.2 g/mol
Density: 1.03 g/cm³ (20 °C)
MDL Number: MFCD00009117
CAS Number: 94-08-6
EINECS: 202-301-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR