1,2-Dihydroxybenzene-3,5-disulfonic acid disodium salt monohydrate

Synonyms: Catechol-3,5-disulphonic acid disodium salt monohydrate, 4,5-Dihydroxy-1,3-benzenedisulphonic acid disodium salt monohydrate, Tiron monohydrate, 3,5-Pyrocatecholdisulphonic acid disodium salt monohydrate, Disodium 4,5-dihydroxybenzene-1,3-disulphonate monohydrate

172553-25G 172553-100G
220039-603EA 35.85 USD
220039-603 220039-602
1,2-Dihydroxybenzene-3,5-disulfonic acid disodium salt monohydrate
1,2-Dihydroxybenzene-3,5-disulphonic acid disodium salt monohydrate
Formula: C₆H₄Na₂O₈S₂·1H₂O
MW: 332.22 g/mol
MDL Number: MFCD00149531
CAS Number: 270573-71-2
EINECS: 205-741-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR